missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Heptyl isothiocyanate, 96%

Manufacturer:  Alfa Aesar™ L1086614

 View more versions of this product

Catalog No. AAL1086614



Specifications

Specifications

1-Heptyl isothiocyanate
Pungent
25g
C8H15NS
UN2922
heptyl isothiocyanate, heptane, 1-isothiocyanato, n-heptyl isothiocyanate, 1-heptyl isothiocyanate, heptylisothiocyanate, n-heptylisothiocyanate, heptane,1-isothiocyanato, acmc-20aks1, 1-dihydroxyboryl butane
CCCCCCCN=C=S
157.275
157.28
106°C (223°F)
233°C
4426-83-9
MFCD00041138
1754818
LIPUQNPCPLDDBO-UHFFFAOYSA-N
1-isothiocyanatoheptane
78152
96%

Chemical Identifiers

4426-83-9
157.275
LIPUQNPCPLDDBO-UHFFFAOYSA-N
78152
CCCCCCCN=C=S
C8H15NS
MFCD00041138
heptyl isothiocyanate, heptane, 1-isothiocyanato, n-heptyl isothiocyanate, 1-heptyl isothiocyanate, heptylisothiocyanate, n-heptylisothiocyanate, heptane,1-isothiocyanato, acmc-20aks1, 1-dihydroxyboryl butane
1-isothiocyanatoheptane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA

Documents