Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Heptyl isocyanate, 98%

Chemical Identifiers

Chemical Name or Material 1-Heptyl isocyanate
CAS 4747-81-3
Molecular Formula CH3(CH2)6NCO
Formula Weight 141.21
MDL Number MFCD00144892
UN Number UN2206
Beilstein 1754817
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2179314

  SDS
alfa aesar™
B2179314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.21
Chemical Name or Material 1-Heptyl isocyanate
Assay 98%
Odor Pungent
Molecular Formula CH3(CH2)6NCO
MDL Number MFCD00144892
CAS 4747-81-3
UN Number UN2206
Beilstein 1754817
Health Hazard 1 Danger
Boiling Point 184°C
Health Hazard 2 GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 62°C (143°F)
Health Hazard 3 GHS P Statement
P210-P284a-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P210-P284a-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.