Please login to your online account to display your discounted pricing

1-Heptene 98%, ACROS Organics™

$154.14 - $154.14

Chemical Identifiers

Chemical Name or Material 1-Heptene
CAS 592-76-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H14
Linear Formula CH3(CH2)4CH=CH2
Formula Weight 98.19
MDL Number MFCD00009531
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC120390250

  SDS
thermo fisher scientific
AC120390250
Glass bottle 25mL Each for $154.14

AC120391000

  SDS
thermo fisher scientific
AC120391000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 98.19
Quantity 25mL
Color Clear colorless
Beilstein 01, 219
Density 0.697g/mL
CAS Max % 100.0
Linear Formula CH3(CH2)4CH=CH2
Refractive Index 1.3990 to 1.4010 (20°C, 589nm)
CAS 592-76-7
MDL Number MFCD00009531
Molecular Formula C7H14
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Assay Percent Range 98%
Chemical Name or Material 1-Heptene
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.