Learn More
1-Heptene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L11487AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Heptene | |
∼-119°C | |
93°C to 94°C | |
C7H14 | |
CH3(CH2)4CH=CH2 | |
100 mL | |
1098332 | |
ZGEGCLOFRBLKSE-UHFFFAOYSA-N | |
hept-1-ene | |
11610 | |
≥98% |
592-76-7 | |
0.697 | |
−8°C (17°F) | |
1.4 | |
MFCD00009531 | |
UN2278 | |
1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum | |
CCCCCC=C | |
98.189 | |
98.19 |
Chemical Identifiers
592-76-7 | |
98.189 | |
ZGEGCLOFRBLKSE-UHFFFAOYSA-N | |
11610 | |
CCCCCC=C |
C7H14 | |
MFCD00009531 | |
1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum | |
hept-1-ene |
Safety and Handling
GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-HEPTENE
EINECSNumber : 209-767-8
RTECSNumber : MJ8840000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only