missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hepten-3-ol, 98%

Manufacturer:  Alfa Aesar™ A1465118

 View more versions of this product

Catalog No. AAA1465118



Specifications

Specifications

1-Hepten-3-ol
154°C to 156°C
4938-52-7
MFCD00021940
1720340
PZKFYTOLVRCMOA-UHFFFAOYSA-N
hept-1-en-3-ol
21057
98%
54°C (129°F)
50g
C7H14O
UN1987
1-hepten-3-ol, heptene-1-ol-3, butyl vinyl carbinol, acmc-1aoyf, 1-hepten-3-ol, 1-hepten-3-ol, analytical standard, 1-hepten-3-ol, fg, 1-hepten-3-ol, purum gc
CCCCC(C=C)O
114.188
114.18

Chemical Identifiers

4938-52-7
114.188
PZKFYTOLVRCMOA-UHFFFAOYSA-N
21057
CCCCC(C=C)O
C7H14O
MFCD00021940
1-hepten-3-ol, heptene-1-ol-3, butyl vinyl carbinol, acmc-1aoyf, 1-hepten-3-ol, 1-hepten-3-ol, analytical standard, 1-hepten-3-ol, fg, 1-hepten-3-ol, purum gc
hept-1-en-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : TSCA

Documents