Please login to your online account to display your discounted pricing

1-Heptanesulfonic Acid Sodium Salt, Monohydrate (HPLC Grade), ACROS Organics™


Quantity 100g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  411271000

 View more versions of this product

Catalog No. AC411271000

  • / Each
 

Description & Specifications

Chemical Identifiers

Absorbance (2.7 g in 100 ml water) (pH 3)
Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt Monohydrate
Color White
Melting Point >300°C
Quantity 100g
CAS 207300-90-1
Absorbance Conditions (2.7g in 100mL water) (pH 3)
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Loss on Drying 7.4% to 9% (100°C, 3 hrs) (vacuum)
Packaging Glass Bottle
Physical Form Powder or Flakes
Molecular Formula C7H15NaO3S·H2O
Linear Formula CH3(CH2)6SO3Na·H2O
Solubility Solubility in water: Soluble
Formula Weight 220.27

Specifications

Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt Monohydrate
CAS 207300-90-1
Molecular Formula C7H15NaO3S·H2O
Linear Formula CH3(CH2)6SO3Na·H2O
Formula Weight 220.27
Accessories

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents