Please login to your online account to display your discounted pricing

1-Heptanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$239.42 - $3,457.92

Chemical Identifiers

Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt
CAS 22767-50-6
Purity Grade HPLC Grade
Molecular Formula CH3(CH2)6SO3Na
Formula Weight 202.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002040

 
avantor performance
217305
Wide Mouth Amber Glass Bottle 25g Each for $239.42

02002041

 
avantor performance
217301
Wide Mouth Amber Glass Bottle 500g Each for $3,457.92
Description & Specifications

Specifications

Formula Weight 202.26
Packaging Wide Mouth Amber Glass Bottle
Purity Grade HPLC Grade
Quantity 25g
Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt
CAS 22767-50-6
Molecular Formula CH3(CH2)6SO3Na
Specific Gravity 1L = 0.00kg

    For Ion-Pair Chromatography of Basic Compounds; For Laboratory,Research,or Manufacturing Use