Please login to your online account to display your discounted pricing

1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC396681000

 View more versions of this product

Catalog No. AC396681000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0% (alkal. after ion exchange) on dry substance
Physical Form Crystalline Powder
Formula Weight 202.25
Quantity 100g
Color White
Purity Grade Ion pair chromatography
CAS Max % 100.0
CAS Min % 99.0
Absorbance 0.02 max. (C=0.5 M in water) at 260nm
Linear Formula CH3(CH2)6SO3Na
Infrared Spectrum Authentic
MDL Number MFCD00007543
Name Note Anhydrous
Chemical Name or Material 1-Heptanesulfonic acid, sodium salt
CAS 22767-50-6
Health Hazard 1 Warning
Loss on Drying 2% max. (150°C)
Packaging Glass bottle
Suitability (HPLC, gradient) Passes
Molecular Formula C7H15NaO3S
Additional Information Membrane filtration test: Passes Test on 0.5M sol
Assay Percent Range 99+%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 1-Heptanesulfonic acid, sodium salt
Name Note Anhydrous
CAS 22767-50-6
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Ion pair chromatography
Assay ≥99.0% (alkal. after ion exchange) on dry substance
Assay Percent Range 99+%
Molecular Formula C7H15NaO3S
Linear Formula CH3(CH2)6SO3Na
Formula Weight 202.25
MDL Number MFCD00007543
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents