missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Heptanesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 206131000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Heptanesulfonic acid, sodium salt | |
White | |
Authentic | |
Glass Bottle | |
CH3(CH2)6SO3Na | |
04, IV, 57 | |
Solubility in water: Soluble | |
CCCCCCCS(=O)(=O)[O-].[Na+] | |
202.244 | |
202.25 | |
Crystalline Powder or Flakes |
22767-50-6 | |
9% max. (100°C, 3 hrs) (vacuum) | |
98% | |
C7H15NaO3S | |
100 g | |
sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate | |
REFMEZARFCPESH-UHFFFAOYSA-M | |
sodium;heptane-1-sulfonate | |
23672332 | |
≥97.5% |
Chemical Identifiers
22767-50-6 | |
sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate | |
sodium;heptane-1-sulfonate |
REFMEZARFCPESH-UHFFFAOYSA-M | |
23672332 | |
CCCCCCCS(=O)(=O)[O-].[Na+] |
Safety and Handling
Warning
RUO – Research Use Only