Please login to your online account to display your discounted pricing

1-Heptadecene, 97%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338330050

 View more versions of this product

Catalog No. AC338330050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 238.46
Quantity 5mL
Color Clear pale yellow
EINECS Number 229-825-6
TSCA TSCA
Density 0.7900g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Linear Formula CH2CH(CH2)14CH3
MDL Number MFCD00009000
CAS 6765-39-5
Chemical Name or Material 1-Heptadecene
Molecular Formula C17H34

Specifications

Chemical Name or Material 1-Heptadecene
Name Note 97%
CAS 6765-39-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C17H34
Linear Formula CH2CH(CH2)14CH3
Formula Weight 238.46
MDL Number MFCD00009000