missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Heptadecene, 97%, ACROS Organics™

Chemical Identifiers

CAS 6765-39-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C17H34
Linear Formula CH2CH(CH2)14CH3
Formula Weight 238.46
MDL Number MFCD00009000
Chemical Name or Material 1-Heptadecene, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC338330050 SDS Thermo Fisher Scientific
338330050
Glass bottle 5mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6765-39-5
100.0
C17H34
238.46
1-Heptadecene, 97%
96.0
97%
CH2CH(CH2)14CH3
MFCD00009000
≥96.0%

Specifications

Clear pale yellow
6765-39-5
100.0
Authentic
Liquid
1.4430 to 1.4450 (20°C, 589nm)
CH2CH(CH2)14CH3
MFCD00009000
≥96.0%
5mL
96.0
0.7900g/mL
Glass bottle
97%
C17H34
238.46
1-Heptadecene, 97%