Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Heptadecanol

$89.59 - $306.31

Chemical Identifiers

Chemical Name or Material 1-Heptadecanol
CAS 1454-85-9
Molecular Formula C17H36O
Linear Formula CH3(CH2)16OH
Formula Weight 256.47
MDL Number MFCD00002822
Beilstein 1702195
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2040414

  SDS
alfa aesar™
L2040414
25g Each for $306.31

AAL2040406

  SDS
alfa aesar™
L2040406
5g Each for $89.59
Description & Specifications

Specifications

Formula Weight 256.47
Chemical Name or Material 1-Heptadecanol
MDL Number MFCD00002822
CAS 1454-85-9
TSCA TSCA
Linear Formula CH3(CH2)16OH
Beilstein 1702195
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MI3900000
Flash Point 155°C (311°F)
Boiling Point 308° to 309°C
Melting Point 52° to 56°C
Molecular Formula C17H36O
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.