Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-Glycerol, polymer-supported, 0.8-1.1 mmol/g on Merrifield resin

Chemical Identifiers

Chemical Name or Material (+/-)-1-Glycerol
Name Note polymer-supported, 0.8 to 1.1mmol/g on Merrifield resin
MDL Number MFCD03454040
Synonym 1,2-Propanediol on Merrifield resin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1946806

  SDS
alfa aesar™
L1946806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1946814

  SDS
alfa aesar™
L1946814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD03454040
Name Note polymer-supported, 0.8 to 1.1mmol/g on Merrifield resin
Quantity 5g
Chemical Name or Material (+/-)-1-Glycerol
Synonym 1,2-Propanediol on Merrifield resin