Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Furfurylpyrrole, 99%

Chemical Identifiers

Chemical Name or Material 1-Furfurylpyrrole
CAS 1438-94-4
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00010080
UN Number UN2810
Synonym 1-(2-Furylmethyl)-1H-pyrrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2310506

 
alfa aesar™
B2310506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2310514

 
alfa aesar™
B2310514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2310522

 
alfa aesar™
B2310522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.18
Chemical Name or Material 1-Furfurylpyrrole
TSCA TSCA
Assay 99%
Molecular Formula C9H9NO
MDL Number MFCD00010080
CAS 1438-94-4
UN Number UN2810
Synonym 1-(2-Furylmethyl)-1H-pyrrole
Health Hazard 1 Warning
Boiling Point 76° to 78°C (1mmHg)
Health Hazard 2 GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Quantity 5g
Flash Point 92°C (199°F)
Health Hazard 3 GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number UX9631000
Safety and Handling

Warning

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.