missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Formylpiperidine, TCI America™

Manufacturer:  TCI America F023425ML

 View more versions of this product

Catalog No. F023425ML



Specifications

Specifications

1-Formylpiperidine
Yellow
2591-86-8
MFCD00006483
n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czg
C1CCN(CC1)C=O
113.16
CHEBI:42546
Clear Liquid at 20°C
222°C
25mL
C6H11NO
2810
FEWLNYSYJNLUOO-UHFFFAOYSA-N
piperidine-1-carbaldehyde
17429
113.16
≥98.0% (GC)

Chemical Identifiers

2591-86-8
113.16
FEWLNYSYJNLUOO-UHFFFAOYSA-N
17429
piperidine-1-carbaldehyde
C6H11NO
MFCD00006483
n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czg
CHEBI:42546
C1CCN(CC1)C=O
Safety and Handling

Safety and Handling

RTECSNumber : TN0380000

TSCA : Yes

Documents