Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Formylhomopiperazine, 99%


Quantity 1g

Manufacturer: alfa aesar™  L17575-03

 View more versions of this product

Catalog No. AAL1757503

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Boiling Point 105°C (0.6mmHg)
Quantity 1g
Chemical Name or Material 1-Formylhomopiperazine
CAS 29053-62-1
Assay 99%
Molecular Formula C6H12N2O
Formula Weight 128.17g/mol
MDL Number MFCD01318149
UN Number UN2735
Beilstein 508208
Synonym 1-Formylhexahydro-1,4-diazepine; Hexahydro-1,4-diazepine-1-carboxaldehyde
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Specifications

Chemical Name or Material 1-Formylhomopiperazine
CAS 29053-62-1
Molecular Formula C6H12N2O
Formula Weight 128.17g/mol
MDL Number MFCD01318149
UN Number UN2735
Beilstein 508208
Synonym 1-Formylhexahydro-1,4-diazepine; Hexahydro-1,4-diazepine-1-carboxaldehyde
Accessories

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.