missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Fmoc-amino)cyclohexanecarboxylic acid, 98+%

$95.30 - $360.51

Chemical Identifiers

CAS 162648-54-6
Molecular Formula C22H23NO4
Formula Weight 365.43
MDL Number MFCD00273464
Beilstein 7887646
Synonym 1-(9-Fluorenylmethoxycarbonylamino)cyclohexanecarboxylic acid
Chemical Name or Material 1-(Fmoc-amino)cyclohexanecarboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1981003 SDS Alfa Aesar™
L1981003
1g Each for $95.30
AAL1981006 SDS Alfa Aesar™
L1981006
5g Each for $360.51
Specifications

Description

Applications
1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

162648-54-6
365.43
7887646
1-(Fmoc-amino)cyclohexanecarboxylic acid
C22H23NO4
MFCD00273464
1-(9-Fluorenylmethoxycarbonylamino)cyclohexanecarboxylic acid

Specifications

185° to 187°C
162648-54-6
365.43
7887646
1-(Fmoc-amino)cyclohexanecarboxylic acid
1g
C22H23NO4
MFCD00273464
1-(9-Fluorenylmethoxycarbonylamino)cyclohexanecarboxylic acid
≥98%

Safety and Handling

Recommended Storage : 2° to 8°C