missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoropyridinium Trifluoromethanesulfonate [Fluorinating Reagent], TCI America™

Manufacturer:  TCI America F032725G

 View more versions of this product

Catalog No. F032725G



Specifications

Specifications

1-Fluoropyridinium Trifluoromethanesulfonate [Fluorinating Reagent]
183°C
107263-95-6
MFCD00013458
1-fluoropyridinium triflate, n-fluoropyridinium triflate, n-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridin-1-ium; trifluoromethanesulfonate, acmc-209shu, ksc182s3j, 1-fluoropyridin-1-ium triflate, 1-fluoropyridiniumtriflate, 1-fluoropyridinium triflate
C1=CC=[N+](C=C1)F.C(F)(F)(F)S(=O)(=O)[O-]
247.164
247.16
≥96.0% (T)
White-Yellow
25g
C6H5F4NO3S
1759
JFZMMCYRTJBQQI-UHFFFAOYSA-M
1-fluoropyridin-1-ium;trifluoromethanesulfonate
2724576
Crystal-Powder at 20°C

Chemical Identifiers

107263-95-6
247.164
JFZMMCYRTJBQQI-UHFFFAOYSA-M
2724576
C1=CC=[N+](C=C1)F.C(F)(F)(F)S(=O)(=O)[O-]
C6H5F4NO3S
MFCD00013458
1-fluoropyridinium triflate, n-fluoropyridinium triflate, n-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridin-1-ium; trifluoromethanesulfonate, acmc-209shu, ksc182s3j, 1-fluoropyridin-1-ium triflate, 1-fluoropyridiniumtriflate, 1-fluoropyridinium triflate
1-fluoropyridin-1-ium;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents