missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoropyridinium Tetrafluoroborate [Fluorinating Reagent], TCI America™

Manufacturer:  TCI America F034325G

 View more versions of this product

Catalog No. F034325G



Specifications

Specifications

1-Fluoropyridinium Tetrafluoroborate [Fluorinating Reagent]
194°C
107264-09-5
MFCD00153176
1-fluoropyridinium tetrafluoroborate, n-fluoropyridinium tetrafluoroborate, 1-fluoropyridin-1-ium tetrafluoroborate, acmc-209tq2, ksc496g3f, 1-fluoropyridinium tetrafluoroborate
[B-](F)(F)(F)F.C1=CC=[N+](C=C1)F
184.904
184.90
≥95.0% (T)
White-Yellow
25g
C5H5BF5N
1759
PIGQKECRKGMXRG-UHFFFAOYSA-N
1-fluoropyridin-1-ium;tetrafluoroborate
2733303
Crystal-Powder at 20°C

Chemical Identifiers

107264-09-5
184.904
PIGQKECRKGMXRG-UHFFFAOYSA-N
2733303
[B-](F)(F)(F)F.C1=CC=[N+](C=C1)F
C5H5BF5N
MFCD00153176
1-fluoropyridinium tetrafluoroborate, n-fluoropyridinium tetrafluoroborate, 1-fluoropyridin-1-ium tetrafluoroborate, acmc-209tq2, ksc496g3f, 1-fluoropyridinium tetrafluoroborate
1-fluoropyridin-1-ium;tetrafluoroborate
Safety and Handling

Safety and Handling

TSCA : No

Documents