missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoronaphthalene, TCI America™

Manufacturer:  TCI America F02125G

 View more versions of this product

Catalog No. F02125G



Specifications

Specifications

1-Fluoronaphthalene
Yellow
321-38-0
MFCD00003873
CWLKTJOTWITYSI-UHFFFAOYSA-N
1-fluoronaphthalene
9450
Clear Liquid at 20°C
212°C
5g
C10H7F
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
C1=CC=C2C(=C1)C=CC=C2F
146.164
146.16
≥98.0% (GC)

Chemical Identifiers

321-38-0
146.164
CWLKTJOTWITYSI-UHFFFAOYSA-N
9450
C1=CC=C2C(=C1)C=CC=C2F
C10H7F
MFCD00003873
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
1-fluoronaphthalene
Safety and Handling

Safety and Handling

RTECSNumber : QJ7100000

TSCA : Yes

Documents