Learn More
1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals
$64.03 - $576.05
Chemical Identifiers
CAS | 321-38-0 |
---|---|
Molecular Formula | C10H7F |
Molecular Weight (g/mol) | 146.164 |
MDL Number | MFCD00003873 |
InChI Key | CWLKTJOTWITYSI-UHFFFAOYSA-N |
Synonym | fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene |
PubChem CID | 9450 |
IUPAC Name | 1-fluoronaphthalene |
SMILES | C1=CC=C2C(=C1)C=CC=C2F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1837806
|
Thermo Scientific Chemicals
A1837806 |
5 g |
Each for $64.03
|
|
|||||
AAA1837814
|
Thermo Scientific Chemicals
A1837814 |
25 g |
Each for $206.15
|
|
|||||
AAA1837822
|
Thermo Scientific Chemicals
A1837822 |
100 g |
Each for $576.05
|
|
|||||
Description
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
321-38-0 | |
146.164 | |
CWLKTJOTWITYSI-UHFFFAOYSA-N | |
9450 | |
C1=CC=C2C(=C1)C=CC=C2F |
C10H7F | |
MFCD00003873 | |
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene | |
1-fluoronaphthalene |
Specifications
321-38-0 | |
1.331 | |
65°C (149°F) | |
1.593 | |
5 g | |
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene | |
CWLKTJOTWITYSI-UHFFFAOYSA-N | |
1-fluoronaphthalene | |
9450 | |
98% |
-10°C | |
215°C to 217°C | |
C10H7F | |
MFCD00003873 | |
1906413 | |
Not miscible or difficult to mix in water. | |
C1=CC=C2C(=C1)C=CC=C2F | |
146.164 | |
146.17 | |
1-Fluoronaphthalene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 206-287-0
RTECSNumber : QJ7100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only