missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1837814

 View more versions of this product

Catalog No. AAA1837814



Description

Description

1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

1-Fluoronaphthalene
-10°C
215°C to 217°C
C10H7F
MFCD00003873
1906413
Not miscible or difficult to mix in water.
C1=CC=C2C(=C1)C=CC=C2F
146.164
146.17
321-38-0
1.331
65°C (149°F)
1.593
25 g
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
CWLKTJOTWITYSI-UHFFFAOYSA-N
1-fluoronaphthalene
9450
98%

Chemical Identifiers

321-38-0
146.164
CWLKTJOTWITYSI-UHFFFAOYSA-N
9450
C1=CC=C2C(=C1)C=CC=C2F
C10H7F
MFCD00003873
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
1-fluoronaphthalene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 206-287-0

RTECSNumber : QJ7100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only