Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Fluoro-4-(trifluoromethylthio)benzene, 98+%

Chemical Identifiers

Chemical Name or Material 1-Fluoro-4-(trifluoromethylthio)benzene
CAS 940-76-1
Molecular Formula C7H4F4S
Formula Weight 196.17
MDL Number MFCD00040978
UN Number UN1993
Beilstein 2046333
Synonym 4-Fluorophenyl trifluoromethyl sulfide; alpha,alpha,alpha,4-Tetrafluorothioanisole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1248503

 
alfa aesar™
L1248503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.17
Molecular Formula C7H4F4S
MDL Number MFCD00040978
Chemical Name or Material 1-Fluoro-4-(trifluoromethylthio)benzene
Assay ≥98%
CAS 940-76-1
Synonym 4-Fluorophenyl trifluoromethyl sulfide; alpha,alpha,alpha,4-Tetrafluorothioanisole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point 37°C (99°F)
Odor Unpleasant
Quantity 1g
UN Number UN1993
Beilstein 2046333
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 136° to 138°C
Safety and Handling

Warning

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.