Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Fluoro-4-(trifluoromethoxy)benzene, 99%

Chemical Identifiers

Chemical Name or Material 1-Fluoro-4-(trifluoromethoxy)benzene
CAS 352-67-0
Molecular Formula C7H4F4O
Formula Weight 180.1
MDL Number MFCD00040835
UN Number UN1993
Beilstein 2046330
Synonym 4-(Trifluoromethoxy)fluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2016003

  SDS
alfa aesar™
B2016003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2016006

  SDS
alfa aesar™
B2016006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.1
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Odor Characteristic
UN Number UN1993
Synonym 4-(Trifluoromethoxy)fluorobenzene
Boiling Point 104° to 105°C
Beilstein 2046330
Health Hazard 1 Danger
Quantity 1g
Flash Point 15°C (59°F)
MDL Number MFCD00040835
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 352-67-0
Chemical Name or Material 1-Fluoro-4-(trifluoromethoxy)benzene
Molecular Formula C7H4F4O
Health Hazard 3 GHS P Statement
P210-P280h-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 99%

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P210-P280h-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.