Learn More
1-Fluoro-4-iodobenzene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1200809
Description
4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool dry place. Keep container tightly sealed.
Specifications
1-Fluoro-4-iodobenzene | |
-20°C | |
182°C to 184°C | |
C6H4FI | |
MFCD00001052 | |
1853970 | |
4-fluoroiodobenzene, p-fluoroiodobenzene, benzene, 1-fluoro-4-iodo, 4-iodofluorobenzene, p-iodofluorobenzene, 4-fluoro-1-iodobenzene, 4-fluoro-iodobenzene, 1-fluoro-4-iodo-benzene, 1-iodo-4-fluorobenzene, 4-fluroiodobenzene | |
KGNQDBQYEBMPFZ-UHFFFAOYSA-N | |
1-fluoro-4-iodobenzene | |
9605 | |
99% |
352-34-1 | |
1.925 | |
68°C (154°F) | |
1.583 | |
10 g | |
Light sensitive | |
Insoluble in water. | |
FC1=CC=C(I)C=C1 | |
222.00 | |
222 |
Chemical Identifiers
352-34-1 | |
222.00 | |
KGNQDBQYEBMPFZ-UHFFFAOYSA-N | |
9605 | |
FC1=CC=C(I)C=C1 |
C6H4FI | |
MFCD00001052 | |
4-fluoroiodobenzene, p-fluoroiodobenzene, benzene, 1-fluoro-4-iodo, 4-iodofluorobenzene, p-iodofluorobenzene, 4-fluoro-1-iodobenzene, 4-fluoro-iodobenzene, 1-fluoro-4-iodo-benzene, 1-iodo-4-fluorobenzene, 4-fluroiodobenzene | |
1-fluoro-4-iodobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 206-522-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only