missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoro-3-iodobenzene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1170614

 View more versions of this product

Catalog No. AAA1170614



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Fluoro-3-iodobenzene
-42°C
77°C to 78°C (19 mmHg)
C6H4FI
MFCD00001044
2039305
3-fluoroiodobenzene, m-fluoroiodobenzene, 3-iodofluorobenzene, m-iodofluorobenzene, benzene, 1-fluoro-3-iodo, m-fluorobenzene, 1-fluoro-3-iodo-benzene, 1-iodo-3-fluorobenzene, 3-fluoro-1-iodobenzene, m-fluorjodbenzol
FC1=CC=CC(I)=C1
222.00
222
1121-86-4
1.898
67°C (152°F)
1.584
25 g
Light sensitive
VSKSBSORLCDRHS-UHFFFAOYSA-N
1-fluoro-3-iodobenzene
70725
99%

Chemical Identifiers

1121-86-4
222.00
VSKSBSORLCDRHS-UHFFFAOYSA-N
70725
FC1=CC=CC(I)=C1
C6H4FI
MFCD00001044
3-fluoroiodobenzene, m-fluoroiodobenzene, 3-iodofluorobenzene, m-iodofluorobenzene, benzene, 1-fluoro-3-iodo, m-fluorobenzene, 1-fluoro-3-iodo-benzene, 1-iodo-3-fluorobenzene, 3-fluoro-1-iodobenzene, m-fluorjodbenzol
1-fluoro-3-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 214-339-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only