missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Fluoro-3,5-dimethyl-2-nitrobenzene, 98%

$102.92 - $102.92

Chemical Identifiers

CAS 315-13-9
Molecular Formula C8H8FNO2
Formula Weight 169.16
MDL Number MFCD00052367
Synonym 1,3-Dimethyl-5-fluoro-4-nitrobenzene; 5-Fluoro-4-nitro-m-xylene
Chemical Name or Material 1-Fluoro-3,5-dimethyl-2-nitrobenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2776203 SDS Alfa Aesar™
H2776203
1g Each for $102.92
AAH2776206 SDS Alfa Aesar™
H2776206
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

315-13-9
169.16
1,3-Dimethyl-5-fluoro-4-nitrobenzene; 5-Fluoro-4-nitro-m-xylene
C8H8FNO2
MFCD00052367
1-Fluoro-3,5-dimethyl-2-nitrobenzene

Specifications

54° to 57°C
315-13-9
169.16
1,3-Dimethyl-5-fluoro-4-nitrobenzene; 5-Fluoro-4-nitro-m-xylene
98%
1g
C8H8FNO2
MFCD00052367
1-Fluoro-3,5-dimethyl-2-nitrobenzene

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger