missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoro-3,5-dimethyl-2-nitrobenzene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2776206

 View more versions of this product

Catalog No. AAH2776206

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Fluoro-3,5-dimethyl-2-nitrobenzene
54°C to 57°C
MFCD00052367
benzene, 1-fluoro-3,5-dimethyl-2-nitro, 5-fluoro-4-nitro-m-xylene, 4,6-dimethyl-2-fluoronitrobenzene, 2-fluoro-4,6-dimethyl-nitrobenzene, 1,3-dimethyl-5-fluoro-4-nitrobenzene, benzene,1-fluoro-3,5-dimethyl-2-nitro
CC1=CC(=C(C(=C1)F)[N+](=O)[O-])C
169.155
169.16
315-13-9
C8H8FNO2
5 g
XGLWCYVNZDSDNQ-UHFFFAOYSA-N
1-fluoro-3,5-dimethyl-2-nitrobenzene
21875915
98%

Chemical Identifiers

315-13-9
169.155
XGLWCYVNZDSDNQ-UHFFFAOYSA-N
21875915
CC1=CC(=C(C(=C1)F)[N+](=O)[O-])C
C8H8FNO2
MFCD00052367
benzene, 1-fluoro-3,5-dimethyl-2-nitro, 5-fluoro-4-nitro-m-xylene, 4,6-dimethyl-2-fluoronitrobenzene, 2-fluoro-4,6-dimethyl-nitrobenzene, 1,3-dimethyl-5-fluoro-4-nitrobenzene, benzene,1-fluoro-3,5-dimethyl-2-nitro
1-fluoro-3,5-dimethyl-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only