missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Fluoro-3,5-dimethoxybenzene, 98%

$78.81 - $1,151.38

Chemical Identifiers

CAS 52189-63-6
Molecular Formula C8H9FO2
Formula Weight 156.16
MDL Number MFCD00012445
Synonym 1,3-Dimethoxy-5-fluorobenzene
Chemical Name or Material 1-Fluoro-3,5-dimethoxybenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2481006 SDS Alfa Aesar™
B2481006
5g Each for $78.81
AAB2481014 SDS Alfa Aesar™
B2481014
25g Each for $354.04
AAB2481022 SDS Alfa Aesar™
B2481022
100g Each for $1,151.38
Specifications

Description

Applications
1-Fluoro-3,5-dimethoxybenzene is used in the preparation of 5-fluoro-benzene-1,3-diol using reagents like tetrabutylammnoium iodide and boron trichloirde.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

52189-63-6
156.16
1,3-Dimethoxy-5-fluorobenzene
C8H9FO2
MFCD00012445
1-Fluoro-3,5-dimethoxybenzene

Specifications

52189-63-6
86° to 88°C (7mmHg)
MFCD00012445
91°C (196°F)
1-Fluoro-3,5-dimethoxybenzene
C8H9FO2
156.16
5g
1,3-Dimethoxy-5-fluorobenzene
98%

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning