Learn More
1-Fluoro-3,5-dimethoxybenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2481006
Description
1-Fluoro-3,5-dimethoxybenzene is used in the preparation of 5-fluoro-benzene-1,3-diol using reagents like tetrabutylammnoium iodide and boron trichloirde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Fluoro-3,5-dimethoxybenzene is used in the preparation of 5-fluoro-benzene-1,3-diol using reagents like tetrabutylammnoium iodide and boron trichloirde.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
1-Fluoro-3,5-dimethoxybenzene | |
1.17 | |
91°C (196°F) | |
1.5025 | |
5 g | |
Slightly miscible with water. | |
COC1=CC(OC)=CC(F)=C1 | |
156.16 | |
156.16 |
52189-63-6 | |
86°C to 88°C (7 mmHg) | |
C8H9FO2 | |
MFCD00012445 | |
1,3-dimethoxy-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethoxy, 3,5-dimethoxyfluorobenzene, pubchem3049, acmc-209wz0, 3,5-dimethoxy fluorobenzene, ksc595k6b, 3,5-dimethoxy-1-fluorobenzene, 1-fluoro-3,5-dimethoxy benzene, benzene,1-fluoro-3,5-dimethoxy | |
IWFKMNAEFPEIOY-UHFFFAOYSA-N | |
1-fluoro-3,5-dimethoxybenzene | |
2774257 | |
98% |
Chemical Identifiers
52189-63-6 | |
156.16 | |
IWFKMNAEFPEIOY-UHFFFAOYSA-N | |
2774257 | |
COC1=CC(OC)=CC(F)=C1 |
C8H9FO2 | |
MFCD00012445 | |
1,3-dimethoxy-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethoxy, 3,5-dimethoxyfluorobenzene, pubchem3049, acmc-209wz0, 3,5-dimethoxy fluorobenzene, ksc595k6b, 3,5-dimethoxy-1-fluorobenzene, 1-fluoro-3,5-dimethoxy benzene, benzene,1-fluoro-3,5-dimethoxy | |
1-fluoro-3,5-dimethoxybenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only