Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Fluoro-2-(trifluoromethoxy)benzene, 97+%

Chemical Identifiers

Chemical Name or Material 1-Fluoro-2-(trifluoromethoxy)benzene
CAS 2106-18-5
Molecular Formula C7H4F4O
Formula Weight 180.1
MDL Number MFCD00236322
UN Number UN1993
Synonym 2-(Trifluoromethoxy)fluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2398103

  SDS
alfa aesar™
B2398103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2398106

  SDS
alfa aesar™
B2398106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.1
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1993
Health Hazard 1 Danger
Molecular Formula C7H4F4O
Quantity 1g
Chemical Name or Material 1-Fluoro-2-(trifluoromethoxy)benzene
CAS 2106-18-5
MDL Number MFCD00236322
Health Hazard 2 GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 2-(Trifluoromethoxy)fluorobenzene
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Assay ≥97%
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.