Learn More
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1385230
Description
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.
Solubility
Immiscible in water.
Notes
Avoid excessive heat and light.
Specifications
1-Fluoro-2-nitrobenzene | |
-8°C | |
215°C | |
99% | |
1.5317 | |
250 g | |
607262 | |
Immiscible in water. | |
C1=CC=C(C(=C1)[N+](=O)[O-])F | |
141.101 | |
141.1 | |
Liquid |
1493-27-2 | |
1.34 | |
94°C (201°F) | |
C6H4FNO2 | |
MFCD00007048 | |
UN2810 | |
2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro | |
PWKNBLFSJAVFAB-UHFFFAOYSA-N | |
1-fluoro-2-nitrobenzene | |
73895 | |
99% |
Chemical Identifiers
1493-27-2 | |
141.101 | |
PWKNBLFSJAVFAB-UHFFFAOYSA-N | |
73895 | |
C1=CC=C(C(=C1)[N+](=O)[O-])F |
C6H4FNO2 | |
MFCD00007048 | |
2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro | |
1-fluoro-2-nitrobenzene |
Safety and Handling
GHS H Statement
H301-H311-H331-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
P260-P262-P264b-P270-P280-P301+P310-P302+P350-P305+P351+P338-P310-P314-P330-P332+P313-P361-P363-P501c
H301-H310-H315-H319-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 216-088-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only