missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Fluoro-2-iodobenzene, 99%, stab. with copper

Manufacturer:  Alfa Aesar™ A1241114

 View more versions of this product

Catalog No. AAA1241114


 

Specifications

Specifications

1-Fluoro-2-iodobenzene
Stabilized with copper
348-52-7
71°C (159°F)
C6H4FI
2039770
TYHUGKGZNOULKD-UHFFFAOYSA-N
1-fluoro-2-iodobenzene
67673
188°C to 189°C
-43°C to -40°C
25g
99%
MFCD00001031
2-Fluoroiodobenzene
C1=CC=C(C(=C1)F)I
222.001
222

Chemical Identifiers

348-52-7
222.001
TYHUGKGZNOULKD-UHFFFAOYSA-N
67673
C6H4FI
MFCD00001031
2-Fluoroiodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents