missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoro-2,6-dichloropyridinium Tetrafluoroborate [Fluorinating Reagent], TCI America™

Manufacturer:  TCI America F034425G

 View more versions of this product

Catalog No. F034425G



Specifications

Specifications

1-Fluoro-2,6-dichloropyridinium Tetrafluoroborate [Fluorinating Reagent]
245°C
140623-89-8
MFCD00153157
2,6-Dichloro-1-fluoropyridinium Tetrafluoroborate
[B-](F)(F)(F)F.C1=CC(=[N+](C(=C1)Cl)F)Cl
253.788
253.79
≥96.0% (T)
Yellow
25g
C5H3BCl2F5N
1759
JVYBYXWHPIEUOT-UHFFFAOYSA-N
2,6-dichloro-1-fluoropyridin-1-ium;tetrafluoroborate
2737307
Crystal-Powder at 20°C

Chemical Identifiers

140623-89-8
253.788
JVYBYXWHPIEUOT-UHFFFAOYSA-N
2737307
[B-](F)(F)(F)F.C1=CC(=[N+](C(=C1)Cl)F)Cl
C5H3BCl2F5N
MFCD00153157
2,6-Dichloro-1-fluoropyridinium Tetrafluoroborate
2,6-dichloro-1-fluoropyridin-1-ium;tetrafluoroborate
Safety and Handling

Safety and Handling

TSCA : No

Documents