Learn More
1-Fluoro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
$67.49 - $439.70
Chemical Identifiers
CAS | 70-34-8 |
---|---|
Molecular Formula | C6H3FN2O4 |
Molecular Weight (g/mol) | 186.098 |
MDL Number | MFCD00007056 |
InChI Key | LOTKRQAVGJMPNV-UHFFFAOYSA-N |
Synonym | 2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene |
PubChem CID | 6264 |
ChEBI | CHEBI:53049 |
IUPAC Name | 1-fluoro-2,4-dinitrobenzene |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1187106
|
Thermo Scientific Chemicals
A1187106 |
5 g |
Each for $67.49
|
|
|||||
AAA1187114
|
Thermo Scientific Chemicals
A1187114 |
25 g |
Each for $168.14
|
|
|||||
AAA1187122
|
Thermo Scientific Chemicals
A1187122 |
100 g |
Each for $439.70
|
|
|||||
Description
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.
Solubility
Soluble in chloroform, benzene, ether and propylene glycol.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
70-34-8 | |
186.098 | |
LOTKRQAVGJMPNV-UHFFFAOYSA-N | |
6264 | |
1-fluoro-2,4-dinitrobenzene |
C6H3FN2O4 | |
MFCD00007056 | |
2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene | |
CHEBI:53049 | |
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F |
Specifications
70-34-8 | |
1.482 | |
>110°C (230°F) | |
1.569 | |
5 g | |
398632 | |
2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene | |
LOTKRQAVGJMPNV-UHFFFAOYSA-N | |
1-fluoro-2,4-dinitrobenzene | |
6264 | |
186.1 | |
1-Fluoro-2,4-dinitrobenzene |
24°C to 30°C | |
296°C | |
C6H3FN2O4 | |
MFCD00007056 | |
UN2811 | |
14,4172 | |
Soluble in chloroform,benzene,ether and propylene glycol. | |
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F | |
186.098 | |
CHEBI:53049 | |
99% |
Safety and Handling
GHS H Statement
H301-H311-H331-H318-H341-H315-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.
P260-P262-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P333+P313-P362-P501c
H301+H311+H331-H315-H317-H320-H335-H336-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 200-734-3
RTECSNumber : CZ7800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only