missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Fluoro-2,4-dimethoxybenzene, 97%

$36.09 - $516.61

Chemical Identifiers

CAS 17715-70-7
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00042265
InChI Key QLJNEPOEZGFNEA-UHFFFAOYSA-N
Synonym 2,4-dimethoxy-1-fluorobenzene, 2,4-dimethoxyfluorobenzene, 1,3-dimethoxy-4-fluorobenzene, 4-fluoro-3-methoxyanisole, benzene, 1-fluoro-2,4-dimethoxy, 2,4-dimethoxy-1-fluoro-benzene, acmc-1c4p9, 1-fluoro-2,4-dimethoxy-benzene, qljnepoezgfnea-uhfffaoysa, benzene,1-fluoro-2,4-dimethoxy
PubChem CID 2758413
IUPAC Name 1-fluoro-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)F)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63528MD View Documents Alfa Aesar™
H63528MD
250mg Each for $36.09
 
AAH6352803 View Documents Alfa Aesar™
H6352803
1g Each for $113.49
 
AAH6352806 View Documents Alfa Aesar™
H6352806
5g Each for $516.61
 
Specifications

Chemical Identifiers

17715-70-7
156.156
QLJNEPOEZGFNEA-UHFFFAOYSA-N
2758413
COC1=CC(=C(C=C1)F)OC
C8H9FO2
MFCD00042265
2,4-dimethoxy-1-fluorobenzene, 2,4-dimethoxyfluorobenzene, 1,3-dimethoxy-4-fluorobenzene, 4-fluoro-3-methoxyanisole, benzene, 1-fluoro-2,4-dimethoxy, 2,4-dimethoxy-1-fluoro-benzene, acmc-1c4p9, 1-fluoro-2,4-dimethoxy-benzene, qljnepoezgfnea-uhfffaoysa, benzene,1-fluoro-2,4-dimethoxy
1-fluoro-2,4-dimethoxybenzene

Specifications

1.188g/mL
250mg
C8H9FO2
2,4-dimethoxy-1-fluorobenzene, 2,4-dimethoxyfluorobenzene, 1,3-dimethoxy-4-fluorobenzene, 4-fluoro-3-methoxyanisole, benzene, 1-fluoro-2,4-dimethoxy, 2,4-dimethoxy-1-fluoro-benzene, acmc-1c4p9, 1-fluoro-2,4-dimethoxy-benzene, qljnepoezgfnea-uhfffaoysa, benzene,1-fluoro-2,4-dimethoxy
COC1=CC(=C(C=C1)F)OC
156.156
156.15
97%
Yellow
17715-70-7
MFCD00042265
QLJNEPOEZGFNEA-UHFFFAOYSA-N
1-fluoro-2,4-dimethoxybenzene
2758413
Liquid
1-Fluoro-2,4-dimethoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning