missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoro-2,4,6-trimethylpyridinium Trifluoromethanesulfonate [Fluorinating Reagent], TCI America™

Manufacturer:  TCI America F03285G

 View more versions of this product

Catalog No. F03285G



Specifications

Specifications

1-Fluoro-2,4,6-trimethylpyridinium Trifluoromethanesulfonate [Fluorinating Reagent]
165°C
107264-00-6
MFCD00067525
1-fluoro-2,4,6-trimethylpyridinium triflate, n-fluoro-2,4,6-trimethylpyridinium triflate, 1-fluoro-2,4,6-trimethylpyridin-1-ium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium triflate, 1-fluoro-2,4,6-trimethylpyridinium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium; trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridine, trifluoromethanesulfonic acid, acmc-1bprl, 1-fluoro-2,4,6-trimethylpyridiniumtriflate, 1-fluoro-2,4,6-trimethyl-pyridinium triflate
CC1=CC(=[N+](C(=C1)C)F)C.C(F)(F)(F)S(=O)(=O)[O-]
289.245
289.24
≥95.0% (HPLC,T)
White-Yellow
5g
C9H11F4NO3S
1759
PRIGFEJKMMRJSF-UHFFFAOYSA-M
1-fluoro-2,4,6-trimethylpyridin-1-ium;trifluoromethanesulfonate
2724575
Crystal-Powder at 20°C

Chemical Identifiers

107264-00-6
289.245
PRIGFEJKMMRJSF-UHFFFAOYSA-M
2724575
CC1=CC(=[N+](C(=C1)C)F)C.C(F)(F)(F)S(=O)(=O)[O-]
C9H11F4NO3S
MFCD00067525
1-fluoro-2,4,6-trimethylpyridinium triflate, n-fluoro-2,4,6-trimethylpyridinium triflate, 1-fluoro-2,4,6-trimethylpyridin-1-ium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium triflate, 1-fluoro-2,4,6-trimethylpyridinium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium; trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridine, trifluoromethanesulfonic acid, acmc-1bprl, 1-fluoro-2,4,6-trimethylpyridiniumtriflate, 1-fluoro-2,4,6-trimethyl-pyridinium triflate
1-fluoro-2,4,6-trimethylpyridin-1-ium;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents