Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-Ferrocenylethanol, 99%

$35.22 - $117.74

Chemical Identifiers

Chemical Name or Material (+/-)-1-Ferrocenylethanol
CAS 1277-49-2
Molecular Formula C12H14FeO
Formula Weight 230.09g/mol
MDL Number MFCD00013752
Synonym 1-Hydroxyethylferrocene; alpha-Methylferrocenemethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1968103

 
alfa aesar™
L19681-03
1g Each for $35.22

AAL1968106

 
alfa aesar™
L19681-06
5g Each for $117.74
Description & Specifications

Specifications

Melting Point 77° to 79°C
Quantity 1g
Flash Point >100°C (212°F)
Chemical Name or Material (+/-)-1-Ferrocenylethanol
CAS 1277-49-2
Assay 99%
Molecular Formula C12H14FeO
Formula Weight 230.09g/mol
MDL Number MFCD00013752
Synonym 1-Hydroxyethylferrocene; alpha-Methylferrocenemethanol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number LK0730000

Applications
(+/-)-1-Ferrocenylethanol is a ligand in titanium-catalyzed intermolecular hydroamination of terminal alkynes (Markovnikov vs. anti-Markovnikov addition). Used as a reactant for single-electron transfer living radical polymerization reactions, preparation of ferrocene-modified thiopyrimidines as anticancer agents, preparation of rersible addition-fragmentation chain transfer (RAFT) agent for RAFT polymerization of styrene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.