missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethynylpyrene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3732706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Ethynylpyrene | |
113°C to 116°C | |
C18H10 | |
5 g | |
1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2 | |
C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 | |
226.28 | |
≥98% |
34993-56-1 | |
Yellow | |
MFCD02093933 | |
2365008 | |
VEBUBSLYGRMOSZ-UHFFFAOYSA-N | |
1-ethynylpyrene | |
154905 | |
Powder |
Chemical Identifiers
34993-56-1 | |
226.28 | |
VEBUBSLYGRMOSZ-UHFFFAOYSA-N | |
154905 | |
C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
C18H10 | |
MFCD02093933 | |
1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2 | |
1-ethynylpyrene |
Safety and Handling
TSCA : No
RUO – Research Use Only