missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethynylpyrene, 96%

Manufacturer:  Alfa Aesar™ L1722703

 View more versions of this product

Catalog No. AAL1722703



Description

Description

Applications
1-Ethynylpyrene is used as a pharmaceutical intermediate.

Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

1-Ethynylpyrene
1g
C18H10
2365008
VEBUBSLYGRMOSZ-UHFFFAOYSA-N
1-ethynylpyrene
154905
96%
113°C to 116°C
34993-56-1
MFCD02093933
1-ethynyl pyrene
C#CC1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1
226.278
226.28

Chemical Identifiers

34993-56-1
226.278
VEBUBSLYGRMOSZ-UHFFFAOYSA-N
154905
C#CC1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1
C18H10
MFCD02093933
1-ethynyl pyrene
1-ethynylpyrene
Documents