missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethynylcyclohexylamine, 98%

Manufacturer:  Alfa Aesar™ H5351414

 View more versions of this product

Catalog No. AAH5351414



Description

Description

Applications
1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.

Notes
Air sensitive. Incompatible with oxidizing agents and acids.
Specifications

Specifications

1-Ethynylcyclohexylamine
42°C (108°F)
Air Sensitive
25g
C8H13N
UN2734
GDKOYYDQISQOMH-UHFFFAOYSA-N
1-ethynylcyclohexan-1-amine
121691
98%
0.913
1.482
65°C to 66°C (20mmHg)
30389-18-5
MFCD00001489
1-ethinyl-cyclohexyl-amine
C#CC1(CCCCC1)N
123.199
123.2

Chemical Identifiers

30389-18-5
123.199
GDKOYYDQISQOMH-UHFFFAOYSA-N
121691
C#CC1(CCCCC1)N
C8H13N
MFCD00001489
1-ethinyl-cyclohexyl-amine
1-ethynylcyclohexan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

EINECSNumber : 250-172-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents