missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethynyl-4-(trifluoromethyl)benzene, TCI America™

Manufacturer:  TCI America E06261G

 View more versions of this product

Catalog No. E06261G



Specifications

Specifications

1-Ethynyl-4-(trifluoromethyl)benzene
Yellow
705-31-7
MFCD01861903
1-ethynyl-4-trifluoromethyl benzene, 4'-trifluoromethylphenyl acetylene, 4-trifluoromethyl phenylacetylene, 4-ethynyl-alpha,alpha,alpha-trifluorotoluene, 4-ethynylbenzotrifluoride, 4-ethynyl-a,a,a-trifluorotoluene, 4-ethynyl trifluorotoluene, benzene, 1-ethynyl-4-trifluoromethyl, 1-ethynyl-4-trifluoromethyl-benzene, 4-ethynyl-1-trifluoromethyl benzene
C#CC1=CC=C(C=C1)C(F)(F)F
170.134
170.13
≥98.0% (GC)
60°C
1g
C9H5F3
1993
XTKBMZQCDBHHKY-UHFFFAOYSA-N
1-ethynyl-4-(trifluoromethyl)benzene
4348295
Clear Liquid at 20°C

Chemical Identifiers

705-31-7
170.134
XTKBMZQCDBHHKY-UHFFFAOYSA-N
4348295
C#CC1=CC=C(C=C1)C(F)(F)F
C9H5F3
MFCD01861903
1-ethynyl-4-trifluoromethyl benzene, 4'-trifluoromethylphenyl acetylene, 4-trifluoromethyl phenylacetylene, 4-ethynyl-alpha,alpha,alpha-trifluorotoluene, 4-ethynylbenzotrifluoride, 4-ethynyl-a,a,a-trifluorotoluene, 4-ethynyl trifluorotoluene, benzene, 1-ethynyl-4-trifluoromethyl, 1-ethynyl-4-trifluoromethyl-benzene, 4-ethynyl-1-trifluoromethyl benzene
1-ethynyl-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents