missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethynyl-4-propylbenzene, TCI America™

Manufacturer:  TCI America E075025G

 View more versions of this product

Catalog No. E075025G



Specifications

Specifications

1-Ethynyl-4-propylbenzene
25g
C11H12
4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
CCCC1=CC=C(C=C1)C#C
144.217
144.22
≥96.0% (GC)
Undesignated
62452-73-7
MFCD00173886
UVFFOABHOIMLNB-UHFFFAOYSA-N
1-ethynyl-4-propylbenzene
2775133
Clear Liquid at 20°C

Chemical Identifiers

62452-73-7
144.217
UVFFOABHOIMLNB-UHFFFAOYSA-N
2775133
CCCC1=CC=C(C=C1)C#C
C11H12
MFCD00173886
4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
1-ethynyl-4-propylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents