missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethynyl-4-(phenylethynyl)benzene, TCI America™

Manufacturer:  TCI America E09671G

 View more versions of this product

Catalog No. E09671G



Specifications

Specifications

1-Ethynyl-4-(phenylethynyl)benzene
90°C
92866-00-7
4-Ethynyldiphenylacetylene, 1-(4-Ethynylphenyl)-2-phenylacetylene
C#CC1=CC=C(C=C1)C#CC2=CC=CC=C2
202.256
202.26
≥98.0% (GC)
White
1g
C16H10
WAUKYPQSFIENDE-UHFFFAOYSA-N
1-ethynyl-4-(2-phenylethynyl)benzene
11275700
Crystal-Powder at 20°C

Chemical Identifiers

92866-00-7
202.256
4-Ethynyldiphenylacetylene, 1-(4-Ethynylphenyl)-2-phenylacetylene
1-ethynyl-4-(2-phenylethynyl)benzene
C16H10
WAUKYPQSFIENDE-UHFFFAOYSA-N
11275700
C#CC1=CC=C(C=C1)C#CC2=CC=CC=C2
Safety and Handling

Safety and Handling

TSCA : No

Documents