missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethynyl-4-hexylbenzene, TCI America™

Manufacturer:  TCI America E056425G

 View more versions of this product

Catalog No. E056425G



Specifications

Specifications

1-Ethynyl-4-hexylbenzene
25g
C14H18
1-eth-1-ynyl-4-hexylbenzene, 4-hexyl-1-ethynylbenzene, 4-hexylphenylacetylene, 1-ethynyl-4-hexyl-benzene, 4-ethynyl-1-hexylbenzene, acmc-209pie, 4-n-hexylphenylacetylene, 1-ethynyl-4-n-hexylbenzene
CCCCCCC1=CC=C(C=C1)C#C
186.298
186.30
≥95.0% (GC)
Undesignated
79887-11-9
MFCD00173883
NFPDFDTYANKKIU-UHFFFAOYSA-N
1-ethynyl-4-hexylbenzene
2775129
Clear Liquid at 20°C

Chemical Identifiers

79887-11-9
186.298
NFPDFDTYANKKIU-UHFFFAOYSA-N
2775129
CCCCCCC1=CC=C(C=C1)C#C
C14H18
MFCD00173883
1-eth-1-ynyl-4-hexylbenzene, 4-hexyl-1-ethynylbenzene, 4-hexylphenylacetylene, 1-ethynyl-4-hexyl-benzene, 4-ethynyl-1-hexylbenzene, acmc-209pie, 4-n-hexylphenylacetylene, 1-ethynyl-4-n-hexylbenzene
1-ethynyl-4-hexylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents