missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethynyl-3-fluorobenzene 98.0+%, TCI America™
Supplier: TCI America E13325G
Specifications
1-Ethynyl-3-fluorobenzene | |
Yellow | |
C8H5F | |
5 g | |
3-Fluorophenylacetylene | |
C#CC1=CC(=CC=C1)F | |
120.126 | |
120.13 | |
Liquid |
2561-17-3 | |
40°C | |
MFCD03093798 | |
1993 | |
PTRUTZFCVFUTMW-UHFFFAOYSA-N | |
1-ethynyl-3-fluorobenzene | |
520218 | |
≥98.0% (GC) |
Chemical Identifiers
2561-17-3 | |
120.126 | |
PTRUTZFCVFUTMW-UHFFFAOYSA-N | |
520218 | |
C#CC1=CC(=CC=C1)F |
C8H5F | |
MFCD03093798 | |
3-Fluorophenylacetylene | |
1-ethynyl-3-fluorobenzene |
Safety and Handling
TSCA : No