Please login to your online account to display your discounted pricing

1-ethynyl-2-fluorobenzene 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Ethynyl-2-fluorobenzene
CAS 766-49-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5F
Linear Formula FC6H4C2CH
Formula Weight 120.13
MDL Number MFCD00799540
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380990010

  SDS
thermo fisher scientific
AC380990010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC380992500

  SDS
thermo fisher scientific
AC380992500
Glass Ampule with screw-cap 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 120.13
Color Clear yellow to brownish
Quantity 1g
Density 1.06g/mL
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5240 to 1.5270 (20°C, 589nm)
Linear Formula FC6H4C2CH
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 1-Ethynyl-2-fluorobenzene
Assay Percent Range 97%
Molecular Formula C8H5F
MDL Number MFCD00799540
CAS 766-49-4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.