Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethynyl-2,2,6-trimethylcyclohexanol, (E)+(Z), 97%

$99.43 - $384.82

Chemical Identifiers

Chemical Name or Material 1-Ethynyl-2,2,6-trimethylcyclohexanol
CAS 41613-59-6
Name Note (E)+(Z)
Molecular Formula C11H18O
Formula Weight 166.26
MDL Number MFCD00798600
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2105706

  SDS
alfa aesar™
B2105706
5g Each for $99.43

AAB2105714

  SDS
alfa aesar™
B2105714
25g Each for $384.82
Description & Specifications

Specifications

Formula Weight 166.26
Assay 97%
Quantity 5g
Chemical Name or Material 1-Ethynyl-2,2,6-trimethylcyclohexanol
TSCA TSCA
Name Note (E)+(Z)
Molecular Formula C11H18O
MDL Number MFCD00798600
CAS 41613-59-6