Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Ethylsulfonyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(Ethylsulfonyl)piperazine
CAS 62937-96-6
Formula Weight 178.25
MDL Number MFCD02093528
Synonym 1-(Ethanesulfonyl)piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5550303

 
alfa aesar™
H5550303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5550306

 
alfa aesar™
H5550306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5550314

 
alfa aesar™
H5550314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 69° to 72°C
Formula Weight 178.25
Chemical Name or Material 1-(Ethylsulfonyl)piperazine
Color White to pale brown
Assay 97%
Physical Form Solid
Synonym 1-(Ethanesulfonyl)piperazine
Quantity 1g
MDL Number MFCD02093528
CAS 62937-96-6