missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Ethylsulfonyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5550306

 View more versions of this product

Catalog No. AAH5550306



Specifications

Specifications

1-(Ethylsulfonyl)piperazine
69°C to 72°C
62937-96-6
MFCD02093528
BIYGAOBOLDXNHM-UHFFFAOYSA-N
1-ethylsulfonylpiperazine
415294
Solid
Brown to White
5g
C6H14N2O2S
1-ethanesulfonyl piperazine
CCS(=O)(=O)N1CCNCC1
178.25
178.25
97%

Chemical Identifiers

62937-96-6
178.25
BIYGAOBOLDXNHM-UHFFFAOYSA-N
415294
CCS(=O)(=O)N1CCNCC1
C6H14N2O2S
MFCD02093528
1-ethanesulfonyl piperazine
1-ethylsulfonylpiperazine
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents